siRNA Details
| Gene Symbol: Hcrtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013064.1 | NP_037196.1 | 5, 6 | 1189 | ||
| GenBank mRNA | AF041244.1 | - | 5, 6 | 1189 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hcrtr1| Gene Symbol: Hcrtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013064.1 | NP_037196.1 | 5, 6 | 1189 | ||
| GenBank mRNA | AF041244.1 | - | 5, 6 | 1189 | ||
Set Membership: |
|