siRNA Details
| Gene Symbol: Cd74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013069.2 | NP_037201.1 | 4, 5 | 477 | ||
| XM_006254761.2 | XP_006254823.1 | 4, 5 | 535 | |||
| GenBank mRNA | BC059152.1 | - | 4, 5 | 458 | 6890709 | |
| FQ219430.1 | - | 4, 5 | 460 | |||
| FQ225098.1 | - | 4, 5 | 452 | |||
| FQ225666.1 | - | 2, 3 | 96 | |||
| FQ227357.1 | - | 4, 5 | 454 | |||
| FQ227455.1 | - | 4, 5 | 457 | |||
| FQ227561.1 | - | 4, 5 | 459 | |||
| FQ228116.1 | - | 4, 5 | 460 | |||
| FQ231756.1 | - | 4, 5 | 461 | |||
| FQ232412.1 | - | 4, 5 | 502 | |||
| FQ232423.1 | - | 4, 5 | 491 | |||
| FQ232867.1 | - | 4, 5 | 461 | |||
| FQ232996.1 | - | 4, 5 | 454 | |||
| FQ233568.1 | - | 4, 5 | 460 | |||
| FQ233706.1 | - | 4, 5 | 460 | |||
| FQ234457.1 | - | 4, 5 | 463 | |||
| X13044.1 | - | 4, 5 | 433 | |||
| X14254.1 | - | 4, 5 | 441 | |||

Cd74
Genomic Map