siRNA Details
| Gene Symbol: Pcmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013073.3 | NP_037205.2 | 5 | 417 | ||
| GenBank mRNA | BC088417.1 | - | 5 | 417 | 7123213 | |
| D11475.1 | - | 5 | 277 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcmt1| Gene Symbol: Pcmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013073.3 | NP_037205.2 | 5 | 417 | ||
| GenBank mRNA | BC088417.1 | - | 5 | 417 | 7123213 | |
| D11475.1 | - | 5 | 277 | |||