siRNA Details
| Gene Symbol: Rln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013413.1 | NP_038199.1 | 2 | 484 | ||
| XM_006231196.2 | XP_006231258.1 | 2 | 610 | |||
| GenBank mRNA | J00780.1 | - | 2 | 484 | ||
| V01264.1 | - | 2 | 484 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rln1| Gene Symbol: Rln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013413.1 | NP_038199.1 | 2 | 484 | ||
| XM_006231196.2 | XP_006231258.1 | 2 | 610 | |||
| GenBank mRNA | J00780.1 | - | 2 | 484 | ||
| V01264.1 | - | 2 | 484 | |||