siRNA Details
| Gene Symbol: Hspa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013083.2 | NP_037215.1 | 4 | 614 | ||
| GenBank mRNA | BC062017.1 | - | 4 | 574 | 5621611 | |
| M14050.1 | - | 3 | 490 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspa5| Gene Symbol: Hspa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013083.2 | NP_037215.1 | 4 | 614 | ||
| GenBank mRNA | BC062017.1 | - | 4 | 574 | 5621611 | |
| M14050.1 | - | 3 | 490 | |||