siRNA Details
Gene Symbol: Hspa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013083.2 | NP_037215.1 | 9 | 2114 | ||
GenBank mRNA | BC062017.1 | - | 9 | 2074 | 5621611 | |
CB327067.1 | - | 1 | 463 | |||
FQ232006.1 | - | 2 | 622 | |||
M14050.1 | - | 8 | 1990 |
