siRNA Details
| Gene Symbol: Hspa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013083.2 | NP_037215.1 | 9 | 1876 | ||
| GenBank mRNA | BC062017.1 | - | 9 | 1836 | 5621611 | |
| CB327067.1 | - | 1 | 701 | |||
| FQ232006.1 | - | 2 | 384 | |||
| M14050.1 | - | 8 | 1752 | |||

Hspa5
Genomic Map