siRNA Details
| Gene Symbol: Gys2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF346902.1 | - | 3 | 476 | ||
| J05446.1 | - | 3 | 521 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gys2| Gene Symbol: Gys2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF346902.1 | - | 3 | 476 | ||
| J05446.1 | - | 3 | 521 | |||