siRNA Details
| Gene Symbol: Mc4r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013099.2 | NP_037231.1 | 1 | 944 | ||
| GenBank mRNA | U67863.1 | - | 1 | 683 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mc4r| Gene Symbol: Mc4r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013099.2 | NP_037231.1 | 1 | 944 | ||
| GenBank mRNA | U67863.1 | - | 1 | 683 | ||
Set Membership: |
|