siRNA Details
| Gene Symbol: Hnf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013103.1 | NP_037235.1 | 2, 3 | 572 | ||
| GenBank mRNA | BC081826.1 | - | 2, 3 | 687 | 7109422 | |
| EF121973.1 | - | 2, 3 | 542 | |||
| EF122006.1 | - | 2, 3 | 542 | |||
| X56546.1 | - | 2, 3 | 572 | |||

Hnf1b
Genomic Map