siRNA Details
| Gene Symbol: Bmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013107.1 | NP_037239.1 | 6 | 1356 | ||
| GenBank mRNA | AY184240.1 | - | 6 | 1356 | ||
| X58830.1 | - | 459 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmp6| Gene Symbol: Bmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013107.1 | NP_037239.1 | 6 | 1356 | ||
| GenBank mRNA | AY184240.1 | - | 6 | 1356 | ||
| X58830.1 | - | 459 | ||||