siRNA Details
| Gene Symbol: Gckr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013120.2 | NP_037252.1 | 11 | 1128 | ||
| GenBank mRNA | BC081843.1 | - | 11 | 1128 | 7096237 | |
| KJ026952.1 | - | 11 | 951 | |||
| X68497.1 | - | 11 | 972 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gckr| Gene Symbol: Gckr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013120.2 | NP_037252.1 | 11 | 1128 | ||
| GenBank mRNA | BC081843.1 | - | 11 | 1128 | 7096237 | |
| KJ026952.1 | - | 11 | 951 | |||
| X68497.1 | - | 11 | 972 | |||