siRNA Details
| Gene Symbol: Pparg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145366.1 | NP_001138838.1 | 3 | 317 | ||
| NM_001145367.1 | NP_001138839.1 | 3 | 495 | |||
| NM_013124.3 | NP_037256.1 | 2 | 292 | |||
| XM_006237009.2 | XP_006237071.1 | 3 | 288 | |||
| GenBank mRNA | AB011365.1 | - | 1 | 125 | ||
| AB019561.1 | - | 2 | 206 | |||
| AF156665.1 | - | 3 | 317 | |||
| AF156666.1 | - | 2 | 254 | |||
| AF246457.1 | - | 3 | 245 | |||
| AF246458.1 | - | 3 | 285 | |||
| Y12882.2 | - | 2 | 255 | |||

Pparg
Genomic Map