siRNA Details
| Gene Symbol: Scn5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160162.1 | NP_001153634.1 | 5 | 794 | ||
| NM_013125.2 | NP_037257.1 | 5 | 794 | |||
| XM_006244076.2 | XP_006244138.1 | 5 | 964 | |||
| XM_008766670.1 | XP_008764892.1 | 5 | 1992 | |||
| XM_008766671.1 | XP_008764893.1 | 5 | 1992 | |||
| GenBank mRNA | AF353637.1 | - | 4 | 597 | ||
| M27902.1 | - | 5 | 794 | |||

Scn5a
Genomic Map