siRNA Details
| Gene Symbol: Cd38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013127.1 | NP_037259.1 | 2 | 351 | ||
| XM_006251072.2 | XP_006251134.1 | 2 | 632 | |||
| GenBank mRNA | D29646.1 | - | 2 | 351 | ||
| D30795.1 | - | 2 | 346 | |||
| FQ212353.1 | - | 2 | 356 | |||

Cd38
Genomic Map