siRNA Details
| Gene Symbol: Anxa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013132.1 | NP_037264.1 | 7 | 502 | ||
| XM_006232268.2 | XP_006232330.1 | 7 | 637 | |||
| GenBank mRNA | AF051895.1 | - | 7 | 557 | ||
| BC081855.1 | - | 7 | 555 | 7192977 | ||
| FQ213209.1 | - | 8 | 632 | |||
| FQ214176.1 | - | 7 | 532 | |||
| FQ229127.1 | - | 7 | 490 | |||
| M21730.1 | - | 7 | 502 | |||

Anxa5
Genomic Map