siRNA Details
| Gene Symbol: Anxa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013132.1 | NP_037264.1 | 6 | 385 | ||
| XM_006232268.2 | XP_006232330.1 | 6 | 520 | |||
| GenBank mRNA | AF051895.1 | - | 6 | 440 | ||
| BC081855.1 | - | 6 | 438 | 7192977 | ||
| FQ213209.1 | - | 7 | 515 | |||
| FQ214176.1 | - | 6 | 415 | |||
| FQ229127.1 | - | 6 | 373 | |||
| M21730.1 | - | 6 | 385 | |||

Anxa5
Genomic Map