siRNA Details
| Gene Symbol: Hmgcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013134.2 | NP_037266.2 | 10 | 1087 | ||
| XM_006231826.2 | XP_006231888.1 | 10 | 1495 | |||
| GenBank mRNA | BC064654.1 | - | 10 | 1087 | 5600023 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmgcr| Gene Symbol: Hmgcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013134.2 | NP_037266.2 | 10 | 1087 | ||
| XM_006231826.2 | XP_006231888.1 | 10 | 1495 | |||
| GenBank mRNA | BC064654.1 | - | 10 | 1087 | 5600023 | |