siRNA Details
| Gene Symbol: Hmgcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013134.2 | NP_037266.2 | 15, 16 | 2023 | ||
| XM_006231826.2 | XP_006231888.1 | 15, 16 | 2431 | |||
| GenBank mRNA | BC064654.1 | - | 15, 16 | 2023 | 5600023 | |
| X55286.1 | - | 1, 2 | 44 | |||

Hmgcr
Genomic Map