siRNA Details
| Gene Symbol: Ddr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166022.1 | NP_001159494.1 | 18 | 2818 | ||
| NM_013137.2 | NP_037269.2 | 18 | 2905 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddr1| Gene Symbol: Ddr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166022.1 | NP_001159494.1 | 18 | 2818 | ||
| NM_013137.2 | NP_037269.2 | 18 | 2905 | |||