siRNA Details
| Gene Symbol: Itpr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013138.1 | NP_037270.1 | 25 | 3270 | ||
| XM_008772710.1 | XP_008770932.1 | 26 | 3334 | |||
| GenBank mRNA | GQ233031.1 | - | 25 | 3136 | ||
| L06096.1 | - | 25 | 3270 | |||

Itpr3
Genomic Map