siRNA Details
| Gene Symbol: Clcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013147.1 | NP_037279.1 | 17 | 2128 | ||
| XM_008762873.1 | XP_008761095.1 | 16 | 2942 | |||
| GenBank mRNA | AY112736.1 | - | 16 | 1877 | ||
| AY112737.1 | - | 18 | 2038 | |||
| AY112738.1 | - | 17 | 1959 | |||
| AY112739.1 | - | 16 | 1830 | |||
| AY112740.1 | - | 17 | 1956 | |||
| X62894.1 | - | 17 | 2128 | |||

Clcn1
Genomic Map