siRNA Details
| Gene Symbol: Clcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013147.1 | NP_037279.1 | 2 | 358 | ||
| XM_008762873.1 | XP_008761095.1 | 2 | 1274 | |||
| GenBank mRNA | AY112736.1 | - | 2 | 186 | ||
| AY112737.1 | - | 2 | 186 | |||
| AY112738.1 | - | 2 | 186 | |||
| AY112739.1 | - | 2 | 186 | |||
| AY112740.1 | - | 2 | 186 | |||
| X62894.1 | - | 2 | 358 | |||

Clcn1
Genomic Map