siRNA Details
Gene Symbol: Has2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013153.1 | NP_037285.1 | 4 | 2097 | ||
GenBank mRNA | AF008201.1 | - | 4 | 2097 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Has2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013153.1 | NP_037285.1 | 4 | 2097 | ||
GenBank mRNA | AF008201.1 | - | 4 | 2097 |