siRNA Details
| Gene Symbol: Dbh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013158.2 | NP_037290.2 | 6 | 1113 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dbh| Gene Symbol: Dbh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013158.2 | NP_037290.2 | 6 | 1113 | ||