siRNA Details
| Gene Symbol: Cckbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013165.2 | NP_037297.1 | 2 | 245 | ||
| GenBank mRNA | M99418.1 | - | 2 | 336 | ||
| X79208.1 | - | 2 | 201 | |||
More Information
Other Species Targeted
Set Membership: |
|

Cckbr
Genomic Map