siRNA Details
| Gene Symbol: Itgb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013171.1 | NP_037303.1 | 11 | 1859 | ||
| XM_006242383.2 | XP_006242445.1 | 12 | 1955 | |||
| GenBank mRNA | AF003598.1 | - | 9 | 1358 | ||
| BC098806.1 | - | 4 | 656 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itgb7| Gene Symbol: Itgb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013171.1 | NP_037303.1 | 11 | 1859 | ||
| XM_006242383.2 | XP_006242445.1 | 12 | 1955 | |||
| GenBank mRNA | AF003598.1 | - | 9 | 1358 | ||
| BC098806.1 | - | 4 | 656 | |||