siRNA Details
| Gene Symbol: Syt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033680.2 | NP_001028852.2 | 10, 11 | 1411 | ||
| XM_008765352.1 | XP_008763574.1 | 7, 8 | 1393 | |||
| XM_008765353.1 | XP_008763575.1 | 7, 8 | 1387 | |||
| GenBank mRNA | AJ617615.1 | - | 10, 11 | 1411 | ||
| CB615786.1 | - | 1, 2 | 85 | |||
| CO398869.1 | - | 4, 5 | 471 | |||
| DQ181550.2 | - | 7, 8 | 1079 | |||
| X52772.1 | - | 10, 11 | 1594 | |||

Syt1
Genomic Map