siRNA Details
| Gene Symbol: Scn4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013178.1 | NP_037310.1 | 22, 23 | 4338 | ||
| XM_006247559.2 | XP_006247621.1 | 23, 24 | 4688 | |||
| GenBank mRNA | M26643.1 | - | 22, 23 | 4338 | ||
| Y17153.1 | - | 21, 22 | 3889 | |||

Scn4a
Genomic Map