siRNA Details
| Gene Symbol: Itgb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013180.1 | NP_037312.1 | 17 | 2244 | ||
| GenBank mRNA | U60096.1 | - | 17 | 2244 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itgb4| Gene Symbol: Itgb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013180.1 | NP_037312.1 | 17 | 2244 | ||
| GenBank mRNA | U60096.1 | - | 17 | 2244 | ||