siRNA Details
Gene Symbol: Prkar1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013181.1 | NP_037313.1 | 7 | 698 | ||
XM_008768311.1 | XP_008766533.1 | 7 | 754 | |||
GenBank mRNA | M17086.1 | - | 7 | 698 | ||
U75932.1 | - | 4 | 212 |
