siRNA Details
| Gene Symbol: Prkar1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013181.1 | NP_037313.1 | 4 | 481 | ||
| XM_008768311.1 | XP_008766533.1 | 4 | 537 | |||
| GenBank mRNA | M17086.1 | - | 4 | 481 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkar1a| Gene Symbol: Prkar1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013181.1 | NP_037313.1 | 4 | 481 | ||
| XM_008768311.1 | XP_008766533.1 | 4 | 537 | |||
| GenBank mRNA | M17086.1 | - | 4 | 481 | ||