siRNA Details
| Gene Symbol: Mep1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013183.2 | NP_037315.1 | 11 | 1370 | ||
| XM_006254458.2 | XP_006254520.1 | 10 | 1511 | |||
| XM_006254459.2 | XP_006254521.1 | 11 | 1339 | |||
| GenBank mRNA | BC081833.1 | - | 11 | 1346 | 7108543 | |
| M88601.1 | - | 11 | 1371 | |||
| Gene Symbol: LOC100911405 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006254445.2 | XP_006254507.2 | 4 | 1222 | ||

Mep1b
Genomic Map