siRNA Details
| Gene Symbol: Mep1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013183.2 | NP_037315.1 | 5, 6 | 278 | ||
| XM_006254458.2 | XP_006254520.1 | 4, 5 | 419 | |||
| XM_006254459.2 | XP_006254521.1 | 5, 6 | 247 | |||
| GenBank mRNA | BC081833.1 | - | 5, 6 | 254 | 7108543 | |
| M88601.1 | - | 5, 6 | 279 | |||

Mep1b
Genomic Map