siRNA Details
| Gene Symbol: Hnf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270931.1 | NP_001257860.1 | 3 | 447 | ||
| NM_001270933.1 | NP_001257862.1 | 3 | 447 | |||
| NM_022180.2 | NP_071516.2 | 2 | 407 | |||
| XM_006235515.2 | XP_006235577.1 | 2 | 402 | |||
| XM_006235516.2 | XP_006235578.1 | 2 | 402 | |||
| GenBank mRNA | CK483260.1 | - | 2 | 236 | ||
| D10554.1 | - | 2 | 283 | |||
| EF193390.1 | - | 2 | 228 | |||
| EF193392.1 | - | 2 | 209 | |||
| FJ608816.1 | - | 3 | 447 | |||
| FJ608817.1 | - | 3 | 447 | |||
| FJ608818.1 | - | 3 | 447 | |||
| FM042560.1 | - | 2 | 412 | |||
| X57133.1 | - | 2 | 298 | |||

Hnf4a
Genomic Map