siRNA Details
| Gene Symbol: Plcg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013187.1 | NP_037319.1 | 4, 5 | 596 | ||
| GenBank mRNA | J03806.1 | - | 4, 5 | 596 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plcg1| Gene Symbol: Plcg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013187.1 | NP_037319.1 | 4, 5 | 596 | ||
| GenBank mRNA | J03806.1 | - | 4, 5 | 596 | ||