siRNA Details
| Gene Symbol: Pfkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013190.4 | NP_037322.1 | 18, 19 | 1953 | ||
| XM_008772798.1 | XP_008771020.1 | 19, 20 | 2056 | |||
| GenBank mRNA | BC061791.1 | - | 18, 19 | 1963 | 5598160 | |
| X58865.1 | - | 18, 19 | 1953 | |||

Pfkl
Genomic Map