siRNA Details
| Gene Symbol: Maob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013198.1 | NP_037330.1 | 1 | 78 | ||
| GenBank mRNA | BC078886.1 | - | 1 | 85 | ||
| BC089814.1 | - | 1 | 43 | 7323595 | ||
| FQ210326.1 | - | 1 | 139 | |||
| M23601.1 | - | 1 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Maob| Gene Symbol: Maob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013198.1 | NP_037330.1 | 1 | 78 | ||
| GenBank mRNA | BC078886.1 | - | 1 | 85 | ||
| BC089814.1 | - | 1 | 43 | 7323595 | ||
| FQ210326.1 | - | 1 | 139 | |||
| M23601.1 | - | 1 | 78 | |||