siRNA Details
| Gene Symbol: Cd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175577.3 | NP_783167.2 | 8, 9 | 1460 | ||
| XM_006231021.2 | XP_006231083.1 | 8, 9 | 1567 | |||
| XM_008760219.1 | XP_008758441.1 | 7, 8 | 1263 | |||
| GenBank mRNA | AF488727.1 | - | 8, 9 | 1399 | ||

Cd6
Genomic Map