siRNA Details
Gene Symbol: Cpt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031559.2 | NP_113747.2 | 11 | 1230 | ||
XM_006230695.1 | XP_006230757.1 | 12 | 1340 | |||
GenBank mRNA | BC072522.1 | - | 11 | 1230 | 7105825 | |
L07736.1 | - | 11 | 1284 | |||
U88294.1 | - | 10 | 1182 |
