siRNA Details
Gene Symbol: Cpt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031559.2 | NP_113747.2 | 19 | 2310 | ||
XM_006230695.1 | XP_006230757.1 | 20 | 2420 | |||
GenBank mRNA | BC072522.1 | - | 19 | 2310 | 7105825 | |
L07736.1 | - | 19 | 2364 | |||
U88294.1 | - | 18 | 2262 |
