siRNA Details
Gene Symbol: Cpt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031559.2 | NP_113747.2 | 6 | 694 | ||
XM_006230695.1 | XP_006230757.1 | 7 | 804 | |||
GenBank mRNA | BC072522.1 | - | 6 | 694 | 7105825 | |
L07736.1 | - | 6 | 748 | |||
U88294.1 | - | 5 | 646 |
