siRNA Details
| Gene Symbol: Osbpl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172023.1 | NP_742020.1 | 14, 15 | 1351 | ||
| GenBank mRNA | AB089316.1 | - | 14, 15 | 1351 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osbpl1a| Gene Symbol: Osbpl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172023.1 | NP_742020.1 | 14, 15 | 1351 | ||
| GenBank mRNA | AB089316.1 | - | 14, 15 | 1351 | ||