siRNA Details
Gene Symbol: Osbpl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172023.1 | NP_742020.1 | 7 | 674 | ||
GenBank mRNA | AB089316.1 | - | 7 | 674 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Osbpl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172023.1 | NP_742020.1 | 7 | 674 | ||
GenBank mRNA | AB089316.1 | - | 7 | 674 |