siRNA Details
| Gene Symbol: Fkbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191863.1 | NP_001178792.1 | 3 | 483 | ||
| GenBank mRNA | AF531427.1 | - | 2 | 150 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fkbp4| Gene Symbol: Fkbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191863.1 | NP_001178792.1 | 3 | 483 | ||
| GenBank mRNA | AF531427.1 | - | 2 | 150 | ||