siRNA Details
| Gene Symbol: Homez | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152849.2 | NP_690062.3 | 2 | 346 | ||
| XM_006251930.2 | XP_006251992.2 | 2 | 1196 | |||
| XM_008770663.1 | XP_008768885.1 | 2 | 1193 | |||
| XM_008770664.1 | XP_008768886.1 | 3 | 681 | |||
| GenBank mRNA | AY126016.1 | - | 1 | 264 | ||
| BC084701.1 | - | 2 | 346 | 7133916 | ||
| BC085694.1 | - | 2 | 331 | 7135570 | ||

Homez
Genomic Map