siRNA Details
Gene Symbol: Aurka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153296.2 | NP_695208.2 | 8 | 1071 | ||
XM_006235675.2 | XP_006235737.1 | 9 | 1150 | |||
XM_006235676.2 | XP_006235738.1 | 9 | 1167 | |||
GenBank mRNA | AF537333.1 | - | 8 | 1059 | ||
BC104676.1 | - | 8 | 1071 | 7453115 | ||
FQ227101.1 | - | 9 | 1156 |
