siRNA Details
| Gene Symbol: Cops2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153297.1 | NP_695209.1 | 13 | 1412 | ||
| XM_006234901.2 | XP_006234963.1 | 13 | 1362 | |||
| XM_008762146.1 | XP_008760368.1 | 13 | 1218 | |||
| GenBank mRNA | AB081072.1 | - | 13 | 1412 | ||
| BC061864.1 | - | 13 | 1189 | 5597874 | ||
| FQ210124.1 | - | 13 | 1337 | |||
| FQ227565.1 | - | 3 | 178 | |||

Cops2
Genomic Map