siRNA Details
| Gene Symbol: Fzd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153305.1 | NP_695217.1 | 1 | 1038 | ||
| GenBank mRNA | AF455054.1 | - | 1 | 1038 | ||
| DQ211688.1 | - | 1 | 800 | |||
| DQ211689.1 | - | 1 | 800 | |||
More Information
Set Membership: |
|

Fzd9
Genomic Map