siRNA Details
| Gene Symbol: Rbm45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153306.1 | NP_695218.1 | 3 | 545 | ||
| GenBank mRNA | AB036990.1 | - | 3 | 545 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm45| Gene Symbol: Rbm45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153306.1 | NP_695218.1 | 3 | 545 | ||
| GenBank mRNA | AB036990.1 | - | 3 | 545 | ||